missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl Chloride 93.0+%, TCI America™

Supplier:  TCI America A14165G

 View more versions of this product

Catalog No. A14165G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl Chloride
C14H20ClNO8
5 g
NAYYKQAWUWXLPD-KSTCHIGDSA-N
[(2R,3S,4R,5R,6R)-5-acetamido-3,4-diacetyloxy-6-chlorooxan-2-yl]methyl acetate
11100622
≥93.0% (T)
3068-34-6
MFCD00069776
2-Acetamido-2-deoxy-alpha-D-glucopyranosyl Chloride 3,4,6-Triacetate, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-alpha-D-glucose
CC(=O)NC1C(C(C(OC1Cl)COC(=O)C)OC(=O)C)OC(=O)C
365.763
365.76
Crystalline Powder

Chemical Identifiers

3068-34-6
365.763
NAYYKQAWUWXLPD-KSTCHIGDSA-N
11100622
CC(=O)NC1C(C(C(OC1Cl)COC(=O)C)OC(=O)C)OC(=O)C
C14H20ClNO8
MFCD00069776
2-Acetamido-2-deoxy-alpha-D-glucopyranosyl Chloride 3,4,6-Triacetate, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-alpha-D-glucose
[(2R,3S,4R,5R,6R)-5-acetamido-3,4-diacetyloxy-6-chlorooxan-2-yl]methyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions