missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,9-Dimethyl-5-decyne, 96%

Manufacturer:  Alfa Aesar™ H5341403

 View more versions of this product

Catalog No. AAH5341403



Specifications

Specifications

2,9-Dimethyl-5-decyne
1g
C12H22
MFCD00041611
RCFYJIGZQAOTFV-UHFFFAOYSA-N
2,9-dimethyldec-5-yne
140552
96%
92°C (19mmHg)
19550-56-2
(CH3)2CH(CH2)2C≡C(CH2)2CH(CH3)2
2,9-dimethyl-5-decyne, 2,9-dimethyl-dec-5-yne, acmc-1bu20, 5-decyne, 2,9-dimethyl, rcfyjigzqaotfv-uhfffaoysa
CC(C)CCC#CCCC(C)C
166.308
166.31

Chemical Identifiers

19550-56-2
166.308
RCFYJIGZQAOTFV-UHFFFAOYSA-N
140552
CC(C)CCC#CCCC(C)C
C12H22
MFCD00041611
2,9-dimethyl-5-decyne, 2,9-dimethyl-dec-5-yne, acmc-1bu20, 5-decyne, 2,9-dimethyl, rcfyjigzqaotfv-uhfffaoysa
2,9-dimethyldec-5-yne
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents