Learn More
2,9-Dimethyl-1,10-phenanthroline hemihydrate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1139814
Description
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
Solubility
It is soluble in methanol: 0.1 g/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.
Specifications
2,9-Dimethyl-1,10-phenanthroline hemihydrate | |
160°C to 164°C | |
MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843 | |
153179 | |
unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate | |
IEBXFSLFDFHSRD-UHFFFAOYSA-N | |
2,9-dimethyl-1,10-phenanthroline;hydrate | |
67652146 | |
≥98% |
34302-69-7 | |
C28H26N4O | |
25 g | |
14,6449 | |
It is soluble in methanol: 0.1g/mL. | |
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 | |
434.54 | |
217.27 (208.26 Anhydrous) |
Chemical Identifiers
34302-69-7 | |
434.54 | |
IEBXFSLFDFHSRD-UHFFFAOYSA-N | |
67652146 | |
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1 |
C28H26N4O | |
MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843 | |
unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate | |
2,9-dimethyl-1,10-phenanthroline;hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 207-601-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only