Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(9-Carbazolyl)ethylboronic acid diethanolamine ester, 98%

$84.23 - $84.23

Chemical Identifiers

Chemical Name or Material 2-(9-Carbazolyl)ethylboronic acid diethanolamine ester
CAS 501014-45-5
Molecular Formula C18H21BN2O2
Formula Weight 308.19
MDL Number MFCD03788733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19575MC

  SDS
alfa aesar™
L19575MC
100mg Each for $84.23
Description & Specifications

Specifications

Formula Weight 308.19
Chemical Name or Material 2-(9-Carbazolyl)ethylboronic acid diethanolamine ester
Assay 98%
Molecular Formula C18H21BN2O2
MDL Number MFCD03788733
CAS 501014-45-5
Melting Point 244° to 248°C
Quantity 100mg