Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid, 95%

$96.58 - $334.50

Chemical Identifiers

Chemical Name or Material 2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
CAS 874605-59-1
Molecular Formula C10H10N2O2
Formula Weight 190.2
MDL Number MFCD00269632
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5023303

  SDS
alfa aesar™
H5023303
1g Each for $334.50

AAH50233MD

  SDS
alfa aesar™
H50233MD
250mg Each for $96.58
Description & Specifications

Specifications

Formula Weight 190.2
Chemical Name or Material 2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Assay 95%
Molecular Formula C10H10N2O2
MDL Number MFCD00269632
CAS 874605-59-1
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.