missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,8-Dihydroxyquinoline 98.0+%, TCI America™

$93.81 - $275.19

Chemical Identifiers

CAS 15450-76-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00216696
InChI Key ZXZKYYHTWHJHFT-UHFFFAOYSA-N
Synonym 2,8-Quinolinediol, 8-Hydroxy-2-quinolinone
PubChem CID 97250
ChEBI CHEBI:48988
IUPAC Name 8-hydroxy-1H-quinolin-2-one
SMILES C1=CC2=C(C(=C1)O)NC(=O)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D38195G
View Documents
Tci America
D38195G
5g
Each for $93.81
 
D381925G
View Documents
Tci America
D381925G
25g
Each for $275.19
 
Description

Description

Specifications

Chemical Identifiers

15450-76-7
161.16
ZXZKYYHTWHJHFT-UHFFFAOYSA-N
97250
8-hydroxy-1H-quinolin-2-one
C9H7NO2
MFCD00216696
2,8-Quinolinediol, 8-Hydroxy-2-quinolinone
CHEBI:48988
C1=CC2=C(C(=C1)O)NC(=O)C=C2

Specifications

White-Yellow
5g
C9H7NO2
2,8-Quinolinediol, 8-Hydroxy-2-quinolinone
C1=CC2=C(C(=C1)O)NC(=O)C=C2
161.16
CHEBI:48988
Crystal-Powder at 20°C
2,8-Dihydroxyquinoline
290°C
15450-76-7
MFCD00216696
ZXZKYYHTWHJHFT-UHFFFAOYSA-N
8-hydroxy-1H-quinolin-2-one
97250
161.16
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No

SDS