Please login to your online account to display your discounted pricing

2,7-Naphthalenedisulfonic Acid Disodium Salt 95%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: fisher scientific acros  415232500

 View more versions of this product

Catalog No. AC415232500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 332.26
Quantity 250g
Color Off-white to beige
EINECS Number 216-733-6
TSCA TSCA
Assay 40% (typically) 2, 7-isomer
Packaging Glass bottle
Name Note 95%
CAS 36511-65-6
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Loss on Drying 7% max. (1 g, 105°C, 3 hrs)
Chemical Name or Material Naphthalenedisulfonic acid, disodium salt
Molecular Formula C10H6Na2O6S2
MDL Number MFCD00066365

Specifications

Chemical Name or Material Naphthalenedisulfonic acid, disodium salt
Name Note 95%
CAS 36511-65-6
Assay 40% (typically) 2, 7-isomer
Molecular Formula C10H6Na2O6S2
Formula Weight 332.26
MDL Number MFCD00066365
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents