Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Dinitrofluorene, 98%

$114.82 - $114.82

Chemical Identifiers

Chemical Name or Material 2,7-Dinitrofluorene
CAS 5405-53-8
Molecular Formula C13H8N2O4
Formula Weight 256.22
MDL Number MFCD00001121
Beilstein 2057852
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1764306

  SDS
alfa aesar™
A1764306
5g Each for $114.82
Description & Specifications

Specifications

Formula Weight 256.22
Chemical Name or Material 2,7-Dinitrofluorene
Assay 98%
Molecular Formula C13H8N2O4
MDL Number MFCD00001121
CAS 5405-53-8
Beilstein 2057852
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number LL7175000
Melting Point >300°C (dec.)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.