missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,7-Dinitro-9-fluorenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5517622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,7-Dinitro-9-fluorenone | |
292°C to 295°C | |
C13H6N2O5 | |
100 g | |
HDVGAFBXTXDYIB-UHFFFAOYSA-N | |
2,7-dinitrofluoren-9-one | |
35842 | |
97% |
31551-45-8 | |
Yellow to Orange | |
MFCD00001153 | |
2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro | |
[O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O | |
270.20 | |
270.2 | |
Powder |
Chemical Identifiers
31551-45-8 | |
270.20 | |
HDVGAFBXTXDYIB-UHFFFAOYSA-N | |
35842 | |
[O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O |
C13H6N2O5 | |
MFCD00001153 | |
2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro | |
2,7-dinitrofluoren-9-one |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 250-695-1
RTECSNumber : LL9051000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only