Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Dimethyl-9-fluorenone-4-carboxylic acid, 97%

$287.80 - $1,045.69

Chemical Identifiers

Chemical Name or Material 2,7-Dimethyl-9-fluorenone-4-carboxylic acid
CAS 500536-41-4
Molecular Formula C16H12O3
Formula Weight 252.27
MDL Number MFCD04039780
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2522806

  SDS
alfa aesar™
B2522806
5g Each for $287.80

AAB2522814

  SDS
alfa aesar™
B2522814
25g Each for $1,045.69
Description & Specifications

Specifications

Formula Weight 252.27
Chemical Name or Material 2,7-Dimethyl-9-fluorenone-4-carboxylic acid
MDL Number MFCD04039780
CAS 500536-41-4
Quantity 5g
Molecular Formula C16H12O3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.