Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Dimethoxynaphthalene, 98%

$80.66 - $253.99

Chemical Identifiers

Chemical Name or Material 2,7-Dimethoxynaphthalene
CAS 3469-26-9
Molecular Formula C12H12O2
Formula Weight 188.23
MDL Number MFCD00004064
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2797106

 
alfa aesar™
H2797106
5g Each for $80.66

AAH2797114

 
alfa aesar™
H2797114
25g Each for $253.99
Description & Specifications

Specifications

Formula Weight 188.23
Chemical Name or Material 2,7-Dimethoxynaphthalene
Assay 98%
Molecular Formula C12H12O2
MDL Number MFCD00004064
CAS 3469-26-9
Melting Point 137° to 140°C
Quantity 5g