missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Diiodofluorene 98.0+%, TCI America™
$216.24 - $652.41
Chemical Identifiers
| CAS | 16218-28-3 |
|---|---|
| Molecular Formula | C13H8I2 |
| Molecular Weight (g/mol) | 418.016 |
| MDL Number | MFCD00040153 |
| InChI Key | YCWGCTPMBCOCLT-UHFFFAOYSA-N |
| PubChem CID | 634493 |
| IUPAC Name | 2,7-diiodo-9H-fluorene |
| SMILES | C1C2=C(C=CC(=C2)I)C3=C1C=C(C=C3)I |
Chemical Identifiers
| 16218-28-3 | |
| 418.016 | |
| YCWGCTPMBCOCLT-UHFFFAOYSA-N | |
| 2,7-diiodo-9H-fluorene |
| C13H8I2 | |
| MFCD00040153 | |
| 634493 | |
| C1C2=C(C=CC(=C2)I)C3=C1C=C(C=C3)I |
Specifications
| 16218-28-3 | |
| Yellow | |
| MFCD00040153 | |
| YCWGCTPMBCOCLT-UHFFFAOYSA-N | |
| 2,7-diiodo-9H-fluorene | |
| 634493 | |
| ≥98.0% (GC) | |
| 2,7-Diiodofluorene |
| 218°C | |
| C13H8I2 | |
| 1 g | |
| C1C2=C(C=CC(=C2)I)C3=C1C=C(C=C3)I | |
| 418.016 | |
| 418.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No