Learn More
2,7-Dihydroxynaphthalene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 115040250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 7-Dihydroxynaphthalene | |
96.0 | |
184°C to 189°C | |
Authentic | |
Glass bottle | |
C10H6(OH)2 | |
25 g | |
2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene | |
OC1=CC2=CC(O)=CC=C2C=C1 | |
160.17 | |
160.17 | |
Powder |
582-17-2 | |
100.0 | |
Gray | |
96% min. (GC) | |
C10H8O2 | |
MFCD00004085 | |
06, 985 | |
DFQICHCWIIJABH-UHFFFAOYSA-N | |
naphthalene-2,7-diol | |
11397 | |
97% |
Chemical Identifiers
582-17-2 | |
160.17 | |
DFQICHCWIIJABH-UHFFFAOYSA-N | |
11397 | |
OC1=CC2=CC(O)=CC=C2C=C1 |
C10H8O2 | |
MFCD00004085 | |
2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene | |
naphthalene-2,7-diol |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 209-478-7
RUO – Research Use Only