Learn More
2,7-Dihydroxynaphthalene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1022936
Description
2,7-Naphthalenediol is a reagent used in the preparation of monomers of high carbon materials. Also used in the synthesis of splitomicin analogues. Used as Starting material for the synthesis of sulfonic acids and divinylnaphthalenes. It is also used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,7-Naphthalenediol is a reagent used in the preparation of monomers of high carbon materials. Also used in the synthesis of splitomicin analogues. Used as Starting material for the synthesis of sulfonic acids and divinylnaphthalenes. It is also used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications
2,7-Dihydroxynaphthalene | |
183°C to 188°C | |
MFCD00004085 | |
2042383 | |
Soluble in water. | |
OC1=CC2=CC(O)=CC=C2C=C1 | |
160.17 | |
160.17 |
582-17-2 | |
C10H8O2 | |
500 g | |
2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene | |
DFQICHCWIIJABH-UHFFFAOYSA-N | |
naphthalene-2,7-diol | |
11397 | |
97% |
Chemical Identifiers
582-17-2 | |
160.17 | |
DFQICHCWIIJABH-UHFFFAOYSA-N | |
11397 | |
OC1=CC2=CC(O)=CC=C2C=C1 |
C10H8O2 | |
MFCD00004085 | |
2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene | |
naphthalene-2,7-diol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-478-7
RTECSNumber : QJ4780000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only