missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Dihydroxynaphthalene, 97%

Manufacturer:  Alfa Aesar™ A1022918

 View more versions of this product

Catalog No. AAA1022918



Description

Description

Applications
2,7-Naphthalenediol is a reagent used in the preparation of monomers of high carbon materials. Also used in the synthesis of splitomicin analogues. Used as Starting material for the synthesis of sulfonic acids and divinylnaphthalenes. It is also used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications

Specifications

2,7-Dihydroxynaphthalene
50g
C10H8O2
2042383
DFQICHCWIIJABH-UHFFFAOYSA-N
naphthalene-2,7-diol
11397
97%
183°C to 188°C
582-17-2
MFCD00004085
2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene, naphthalenediol-2,7
C1=CC(=CC2=C1C=CC(=C2)O)O
160.172
160.17

Chemical Identifiers

582-17-2
160.172
DFQICHCWIIJABH-UHFFFAOYSA-N
11397
C1=CC(=CC2=C1C=CC(=C2)O)O
C10H8O2
MFCD00004085
2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene, naphthalenediol-2,7
naphthalene-2,7-diol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QJ4780000

TSCA : TSCA

SDS Documents