missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2',7'-Dichlorofluorescein, ACS reagent, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446930100
Specifications
2', 7'-Dichlorofluorescein | |
C20H10Cl2O5 | |
VFNKZQNIXUFLBC-UHFFFAOYSA-N | |
2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one | |
64944 | |
401.2 | |
Glass bottle | |
10g |
76-54-0 | |
2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one | |
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl | |
401.195 | |
CHEBI:51596 | |
ACS Reagent | |
0.7900g/mL |
Chemical Identifiers
76-54-0 | |
401.195 | |
2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one | |
CHEBI:51596 | |
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl |
C20H10Cl2O5 | |
VFNKZQNIXUFLBC-UHFFFAOYSA-N | |
64944 | |
2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |