missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Dibromonaphthalene, 99%

Manufacturer:  Alfa Aesar™ H56097MD

 View more versions of this product

Catalog No. AAH56097MD



Description

Description

Applications
It is an intermediate in dyestuffs and pharmaceuticals.

Solubility
Soluble in acetonitrile.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

2,7-Dibromonaphthalene
250mg
C10H6Br2
UN3077
2,7-dibromonaphthalene hplc
ODJZWBLNJKNOJK-UHFFFAOYSA-N
2,7-dibromonaphthalene
625355
99%
139°C to 142°C
58556-75-5
MFCD00238580
1862041
Soluble in acetonitrile.
C1=CC(=CC2=C1C=CC(=C2)Br)Br
285.966
285.97

Chemical Identifiers

58556-75-5
285.966
ODJZWBLNJKNOJK-UHFFFAOYSA-N
625355
C1=CC(=CC2=C1C=CC(=C2)Br)Br
C10H6Br2
MFCD00238580
2,7-dibromonaphthalene hplc
2,7-dibromonaphthalene
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents