missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Diaminofluorene dihydrochloride, 97%

Manufacturer:  Alfa Aesar™ L1156803

 View more versions of this product

Catalog No. AAL1156803



Specifications

Specifications

2,7-Diaminofluorene dihydrochloride
>360°C
13548-69-1
MFCD00012535
9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl
269.169
269.18
Odorless
1g
C13H14Cl2N2
14,4156
QJXYCUYLZDQRIN-UHFFFAOYSA-N
9H-fluorene-2,7-diamine;dihydrochloride
11543576
97%

Chemical Identifiers

13548-69-1
269.169
QJXYCUYLZDQRIN-UHFFFAOYSA-N
11543576
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl
C13H14Cl2N2
MFCD00012535
9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride
9H-fluorene-2,7-diamine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H341-H351-H302
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement
P280h
Wear protective gloves/clothing.

Warning

RTECSNumber : LL7050000

Documents