missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,7-Diaminofluorene dihydrochloride, 97%, Thermo Scientific™
$76.93 - $76.93
Chemical Identifiers
CAS | 13548-69-1 |
---|---|
Molecular Formula | C13H14Cl2N2 |
Molecular Weight (g/mol) | 269.169 |
MDL Number | MFCD00012535 |
InChI Key | QJXYCUYLZDQRIN-UHFFFAOYSA-N |
Synonym | 9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride |
PubChem CID | 11543576 |
IUPAC Name | 9H-fluorene-2,7-diamine;dihydrochloride |
SMILES | C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1156803
|
Thermo Scientific Chemicals
L1156803 |
1 g |
Each for $76.93
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13548-69-1 | |
269.169 | |
QJXYCUYLZDQRIN-UHFFFAOYSA-N | |
11543576 | |
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl |
C13H14Cl2N2 | |
MFCD00012535 | |
9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride | |
9H-fluorene-2,7-diamine;dihydrochloride |
Specifications
13548-69-1 | |
Odorless | |
MFCD00012535 | |
Light sensitive | |
9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride | |
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl | |
269.169 | |
269.18 | |
2,7-Diaminofluorene dihydrochloride |
>360°C | |
C13H14Cl2N2 | |
1 g | |
14,4156 | |
QJXYCUYLZDQRIN-UHFFFAOYSA-N | |
9H-fluorene-2,7-diamine;dihydrochloride | |
11543576 | |
97% |
Safety and Handling
GHS H Statement
H341-H351-H302
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H351
EINECSNumber : 236-924-8
RTECSNumber : LL7050000
TSCA : No
Recommended Storage : Ambient temperatures