Learn More
2,7-Diaminofluorene, 97+%, Thermo Scientific Chemicals
$51.37 - $538.51
Chemical Identifiers
CAS | 525-64-4 |
---|---|
Molecular Formula | C13H13ClN2 |
Molecular Weight (g/mol) | 232.71 |
MDL Number | MFCD00001128 |
InChI Key | COOWDVCIUYIBFE-UHFFFAOYSA-N |
Synonym | 2,7-diaminofluorene, 2,7-fluorenediamine, 2,7-diamino-fluorene, 2,7-fluoroenediamine, fluorene-2,7-diamine, fluorene-2,7-diyldiamine, ccris 920, 2,7-diamino-fluoren, 2,7-diaminoflyorene, 2,7-diaminofluoren |
PubChem CID | 10679 |
IUPAC Name | 9H-fluorene-2,7-diamine |
SMILES | [Cl-].NC1=CC=C2C(CC3=CC([NH3+])=CC=C23)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH55402MD
|
Thermo Scientific Chemicals
H55402MD |
250 mg |
Each for $51.37
|
|
|||||
AAH5540203
|
Thermo Scientific Chemicals
H5540203 |
1 g |
Each for $144.64
|
|
|||||
AAH5540206
|
Thermo Scientific Chemicals
H5540206 |
5 g |
Each for $538.51
|
|
|||||
Description
It is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.
Solubility
Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether.
Notes
Store at 4°C. Keep away from oxidizing agents.
Chemical Identifiers
525-64-4 | |
232.71 | |
COOWDVCIUYIBFE-UHFFFAOYSA-N | |
10679 | |
[Cl-].NC1=CC=C2C(CC3=CC([NH3+])=CC=C23)=C1 |
C13H13ClN2 | |
MFCD00001128 | |
2,7-diaminofluorene, 2,7-fluorenediamine, 2,7-diamino-fluorene, 2,7-fluoroenediamine, fluorene-2,7-diamine, fluorene-2,7-diyldiamine, ccris 920, 2,7-diamino-fluoren, 2,7-diaminoflyorene, 2,7-diaminofluoren | |
9H-fluorene-2,7-diamine |
Specifications
525-64-4 | |
C13H13ClN2 | |
250 mg | |
14,4156 | |
Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether. | |
[Cl-].NC1=CC=C2C(CC3=CC([NH3+])=CC=C23)=C1 | |
232.71 | |
196.25 | |
2,7-Diaminofluorene |
160°C to 162°C | |
MFCD00001128 | |
2099859 | |
2,7-diaminofluorene, 2,7-fluorenediamine, 2,7-diamino-fluorene, 2,7-fluoroenediamine, fluorene-2,7-diamine, fluorene-2,7-diyldiamine, ccris 920, 2,7-diamino-fluoren, 2,7-diaminoflyorene, 2,7-diaminofluoren | |
COOWDVCIUYIBFE-UHFFFAOYSA-N | |
9H-fluorene-2,7-diamine | |
10679 | |
≥97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-377-5
RTECSNumber : LL6980000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only