Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,7-Diacetoxynaphthalene, 98%

$32.09 - $32.09

Chemical Identifiers

Chemical Name or Material 2,7-Diacetoxynaphthalene
CAS 22472-26-0
Molecular Formula C14H12O4
Formula Weight 244.25
MDL Number MFCD00036008
Synonym 2,7-Naphthalenediol diacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2158803

  SDS
alfa aesar™
B2158803
1g Each for $32.09
Description & Specifications

Specifications

Formula Weight 244.25
Chemical Name or Material 2,7-Diacetoxynaphthalene
Assay 98%
Molecular Formula C14H12O4
MDL Number MFCD00036008
CAS 22472-26-0
Synonym 2,7-Naphthalenediol diacetate
Melting Point 129° to 132°C
Quantity 1g