missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,7-Bis[N-(1-naphthyl)anilino]-9,9′-spirobi[9H-fluorene], TCI America™

$134.45 - $458.34

Chemical Identifiers

CAS 932739-76-9
Molecular Formula C57H38N2
Molecular Weight (g/mol) 750.945
MDL Number MFCD12022428
InChI Key ZDAWFMCVTXSZTC-UHFFFAOYSA-N
PubChem CID 51358306
IUPAC Name 2-N',7-N'-dinaphthalen-1-yl-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine
SMILES C1=CC=C(C=C1)N(C2=CC3=C(C=C2)C4=C(C35C6=CC=CC=C6C7=CC=CC=C57)C=C(C=C4)N(C8=CC=CC=C8)C9=CC=CC1=CC=CC=C19)C1=CC=CC2=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4875200MG View Documents TCI America
B4875200MG
200mg Each for $134.45
 
B48751G View Documents TCI America
B48751G
1g Each for $458.34
 
Specifications

Chemical Identifiers

932739-76-9
750.945
ZDAWFMCVTXSZTC-UHFFFAOYSA-N
2-N',7-N'-dinaphthalen-1-yl-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine
C57H38N2
MFCD12022428
51358306
C1=CC=C(C=C1)N(C2=CC3=C(C=C2)C4=C(C35C6=CC=CC=C6C7=CC=CC=C57)C=C(C=C4)N(C8=CC=CC=C8)C9=CC=CC1=CC=CC=C19)C1=CC=CC2=CC=CC=C21

Specifications

White-Yellow
200mg
C57H38N2
ZDAWFMCVTXSZTC-UHFFFAOYSA-N
2-N',7-N'-dinaphthalen-1-yl-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine
51358306
Crystal-Powder at 20°C
2,7-Bis[N-(1-naphthyl)anilino]-9,9′-spirobi[9H-fluorene]
284°C
932739-76-9
MFCD12022428
C1=CC=C(C=C1)N(C2=CC3=C(C=C2)C4=C(C35C6=CC=CC=C6C7=CC=CC=C57)C=C(C=C4)N(C8=CC=CC=C8)C9=CC=CC1=CC=CC=C19)C1=CC=CC2=CC=CC=C21
750.945
750.95
≥98.0% (HPLC,N)
Safety and Handling

Safety and Handling

TSCA : No