Learn More
2,6-Pyridinedicarboxylic Acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131885000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 6-Pyridinedicarboxylic acid | |
98.5 | |
248°C to 250°C | |
188°C | |
99% | |
C7H5NO4 | |
MFCD00006299 | |
22, 154 | |
Solubility in water: 5g/L (20°C). Other solubilities: soluble in boiling water,soluble in boiling ethanol | |
C1=CC(=NC(=C1)C(=O)O)C(=O)O | |
167.12 | |
CHEBI:46837 | |
99% |
499-83-2 | |
100.0 | |
White | |
Authentic | |
Plastic bottle | |
(C5H3N)(COOH)2 | |
500 g | |
2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid | |
WJJMNDUMQPNECX-UHFFFAOYSA-N | |
pyridine-2,6-dicarboxylic acid | |
10367 | |
167.12 | |
Crystalline Powder |
Chemical Identifiers
499-83-2 | |
MFCD00006299 | |
2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid | |
CHEBI:46837 | |
C1=CC(=NC(=C1)C(=O)O)C(=O)O |
167.12 | |
WJJMNDUMQPNECX-UHFFFAOYSA-N | |
10367 | |
pyridine-2,6-dicarboxylic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning
EINECSNumber : 207-894-3
RUO – Research Use Only