missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Lutidine, TCI America™

Manufacturer:  TCI America L006725ML

 View more versions of this product

Catalog No. L006725ML



Specifications

Specifications

2,6-Lutidine
Yellow
25mL
C7H9N
1993
OISVCGZHLKNMSJ-UHFFFAOYSA-N
2,6-dimethylpyridine
7937
107.16
≥98.0% (GC)
144°C
-6°C
108-48-5
MFCD00006345
2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene
CC1=NC(=CC=C1)C
107.156
CHEBI:32548
Clear Liquid at 20°C

Chemical Identifiers

108-48-5
107.156
OISVCGZHLKNMSJ-UHFFFAOYSA-N
7937
2,6-dimethylpyridine
C7H9N
MFCD00006345
2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene
CHEBI:32548
CC1=NC(=CC=C1)C
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0712

RTECSNumber : OK9700000

TSCA : Yes

Documents