missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Lutidine N-Oxide 98.0+%, TCI America™
Supplier: TCI America L006925G
Specifications
2,6-Lutidine N-Oxide | |
Yellow | |
C7H9NO | |
25 g | |
LIDGFHXPUOJZMK-UHFFFAOYSA-N | |
2,6-dimethyl-1-oxidopyridin-1-ium | |
14083 | |
≥98.0% (GC) |
1073-23-0 | |
145°C | |
MFCD00039715 | |
2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate | |
CC1=[N+](C(=CC=C1)C)[O-] | |
123.155 | |
123.16 | |
Liquid |
Chemical Identifiers
1073-23-0 | |
123.155 | |
LIDGFHXPUOJZMK-UHFFFAOYSA-N | |
14083 | |
CC1=[N+](C(=CC=C1)C)[O-] |
C7H9NO | |
MFCD00039715 | |
2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate | |
2,6-dimethyl-1-oxidopyridin-1-ium |
Safety and Handling
TSCA : Yes