missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Lutidine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC125190025
Specifications
2, 6-Lutidine | |
33°C | |
Glass bottle | |
0.92 | |
143.0°C to 145.0°C | |
-6.0°C | |
99% | |
98.5 | |
98.5% min. (GC) | |
MFCD00006345 | |
01,626 | |
2,6-dimethyl-pyridine, 2,6-dimethypyridine, 2,6-lutidene, 2,6-lutidine, alpha,alpha'-dimethylpyridine, alpha,alpha'-lutidine, hsdb 79, lutidine, pyridine, 2,6-dimethyl, unii-15fq5d0t3p | |
OISVCGZHLKNMSJ-UHFFFAOYSA-N | |
2,6-dimethylpyridine | |
7937 | |
107.15 | |
99% |
0.9200g/mL | |
Authentic | |
1.4966 to 1.4986 | |
0.2% max. | |
Undesignated | |
2.5L | |
108-48-5 | |
100.0 | |
C7H9N | |
20, 244 | |
15, 5677 | |
Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol and ether,miscible with dimethylformamide and,tetrahydrofuran | |
CC1=NC(=CC=C1)C | |
107.15 | |
CHEBI:32548 | |
Liquid |
Chemical Identifiers
108-48-5 | |
107.15 | |
OISVCGZHLKNMSJ-UHFFFAOYSA-N | |
7937 | |
2,6-dimethylpyridine |
C7H9N | |
MFCD00006345 | |
2,6-dimethyl-pyridine, 2,6-dimethypyridine, 2,6-lutidene, 2,6-lutidine, alpha,alpha'-dimethylpyridine, alpha,alpha'-lutidine, hsdb 79, lutidine, pyridine, 2,6-dimethyl, unii-15fq5d0t3p | |
CHEBI:32548 | |
CC1=NC(=CC=C1)C |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable liquid and vapor.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce
GHS Signal Word: Warning
EINECSNumber : 203-587-3
RTECSNumber : OK9700000
TSCA : TSCA
Recommended Storage : This substance may take up color on standing