Learn More
2,6-Lutidine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A10478AE
Description
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.
Solubility
Miscible with water.
Notes
Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
Specifications
2,6-Lutidine | |
-7°C | |
143°C to 145°C | |
Unpleasant | |
1.497 | |
100 mL | |
105690 | |
14,5616 | |
Miscible with water. | |
CC1=CC=CC(C)=N1 | |
107.16 | |
CHEBI:32548 | |
≥98% |
108-48-5 | |
0.923 | |
33°C (91°F) | |
C7H9N | |
MFCD00006345 | |
UN1993 | |
Hygroscopic | |
2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene | |
OISVCGZHLKNMSJ-UHFFFAOYSA-N | |
2,6-dimethylpyridine | |
7937 | |
107.16 |
Chemical Identifiers
108-48-5 | |
107.16 | |
OISVCGZHLKNMSJ-UHFFFAOYSA-N | |
CHEBI:32548 | |
CC1=CC=CC(C)=N1 |
C7H9N | |
MFCD00006345 | |
7937 | |
2,6-dimethylpyridine |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 203-587-3
RTECSNumber : OK9700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only