Please login to your online account to display your discounted pricing

2,6-Diphenylphenol, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  214740250

 View more versions of this product

Catalog No. AC214740250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystals or Powder
Formula Weight 246.31
Quantity 25g
Color White to off-white
Beilstein 06, III, 3631
TSCA TSCA
Melting Point 98° to 103°C
EINECS Number 219-401-9
CAS Min % 96.0
CAS Max % 100.0
CAS 194618
Infrared Spectrum Authentic
Chemical Name or Material 2, 6-Diphenylphenol
Packaging Glass bottle
Name Note 97%
Molecular Formula C18H14O
Linear Formula (C6H4)2C6H3OH
MDL Number MFCD00009716
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%

Specifications

Chemical Name or Material 2, 6-Diphenylphenol
Name Note 97%
CAS 194618
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C18H14O
Linear Formula (C6H4)2C6H3OH
Formula Weight 246.31
MDL Number MFCD00009716
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents