missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Diphenylisonicotinic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 38947-57-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C18H13NO2
Formula Weight 275.3
MDL Number MFCD00418438
Chemical Name or Material 2, 6-Diphenylisonicotinic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC398980010 SDS Thermo Fisher Scientific
AC398980010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398980050 SDS Thermo Fisher Scientific
AC398980050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

38947-57-8
100.0
C18H13NO2
MFCD00418438
96.0
97%
275.3
2, 6-Diphenylisonicotinic acid

Specifications

275.3
96.0
38947-57-8
C18H13NO2
Glass bottle
1g
100.0
97%
MFCD00418438
2, 6-Diphenylisonicotinic acid

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.