Learn More
2,6-Dinitrobenzonitrile, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1262406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Dinitrobenzonitrile | |
146°C to 150°C | |
MFCD00041246 | |
UN3439 | |
dinitrobenzonitrile, benzonitrile, 2,6-dinitro, 2,6-dinitro-benzonitrile, 2,6-dinitrobenzenecarbonitrile, 2,6-dinitrobenznitrile, pubchem21108, 2.6-dinitrobenzonitril, 2,6 dinitrobenzonitrile, 2,6-dini trobenzonitrile, acmc-1ad1y | |
C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-] | |
193.118 | |
193.12 |
35213-00-4 | |
C7H3N3O4 | |
5 g | |
2215516 | |
ZYDGHQSJZAFMLU-UHFFFAOYSA-N | |
2,6-dinitrobenzonitrile | |
520718 | |
≥98% |
Chemical Identifiers
35213-00-4 | |
193.118 | |
ZYDGHQSJZAFMLU-UHFFFAOYSA-N | |
520718 | |
C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-] |
C7H3N3O4 | |
MFCD00041246 | |
dinitrobenzonitrile, benzonitrile, 2,6-dinitro, 2,6-dinitro-benzonitrile, 2,6-dinitrobenzenecarbonitrile, 2,6-dinitrobenznitrile, pubchem21108, 2.6-dinitrobenzonitril, 2,6 dinitrobenzonitrile, 2,6-dini trobenzonitrile, acmc-1ad1y | |
2,6-dinitrobenzonitrile |
Safety and Handling
GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312+H332-H315-H319-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only