Learn More
2,6-Dinitroaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1212022
Description
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from strong acids, acid chlorides, acid anhydrides.
Specifications
2,6-Dinitroaniline | |
∼136°C (decomposition) | |
MFCD00007148 | |
UN1596 | |
14,3271 | |
Insoluble in water. | |
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-] | |
183.123 | |
183.12 |
606-22-4 | |
C6H5N3O4 | |
100 g | |
2214886 | |
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine | |
QFUSCYRJMXLNRB-UHFFFAOYSA-N | |
2,6-dinitroaniline | |
69070 | |
98% |
Chemical Identifiers
606-22-4 | |
183.123 | |
QFUSCYRJMXLNRB-UHFFFAOYSA-N | |
69070 | |
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-] |
C6H5N3O4 | |
MFCD00007148 | |
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine | |
2,6-dinitroaniline |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROANILINES
EINECSNumber : 210-108-1
RTECSNumber : BX9200000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only