missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dinitroaniline, 98%

Manufacturer:  Alfa Aesar™ A1212014

 View more versions of this product

Catalog No. AAA1212014



Description

Description

Applications
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from strong acids, acid chlorides, acid anhydrides.
Specifications

Specifications

2,6-Dinitroaniline
25g
C6H5N3O4
UN1596
14,3271
QFUSCYRJMXLNRB-UHFFFAOYSA-N
2,6-dinitroaniline
69070
98%
~136°C (decomposition)
606-22-4
MFCD00007148
2214886
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]
183.123
183.12

Chemical Identifiers

606-22-4
183.123
QFUSCYRJMXLNRB-UHFFFAOYSA-N
69070
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]
C6H5N3O4
MFCD00007148
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
2,6-dinitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROANILINES

RTECSNumber : BX9200000

SDS Documents