Please login to your online account to display your discounted pricing

2,6-Dinitroaniline, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dinitroaniline
Name Note 97%
CAS 606-22-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C6H5N3O4
Linear Formula (O2N)2C6H3NH2
Formula Weight 183.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170170100

  SDS
thermo fisher scientific
AC170170100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Assay ≥94% (HPLC)
Formula Weight 183.12
Quantity 10g
Color White to off-white
EINECS Number 210-108-1
TSCA TSCA
Melting Point 135° to 142°C
RTECS Number BX9200000
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 6-Dinitroaniline
Name Note 97%
Assay Percent Range 95%
Molecular Formula C6H5N3O4
Linear Formula (O2N)2C6H3NH2
CAS 606-22-4
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Packaging Glass bottle
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.