missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dinitroaniline, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC170170100

 View more versions of this product

Catalog No. AC170170100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dinitroaniline
Authentic
White
10g
606-22-4
100.0
C6H5N3O4
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]
183.123
183.12
95%
>100°C
Glass bottle
137.0°C to 142.0°C
97%
96.0
95%
(O2N)2C6H3NH2
QFUSCYRJMXLNRB-UHFFFAOYSA-N
2,6-dinitroaniline
69070
Crystalline Powder or Crystals

Chemical Identifiers

606-22-4
183.123
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
2,6-dinitroaniline
C6H5N3O4
QFUSCYRJMXLNRB-UHFFFAOYSA-N
69070
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS P Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-108-1

RTECSNumber : BX9200000

TSCA : TSCA

SDS Documents