missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylpyrazine, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC116691000

 View more versions of this product

Catalog No. AC116691000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 6-Dimethylpyrazine
Pale yellow
100g
108-50-9
100.0
Glass bottle
≥95%
C6H8N2
MFCD00006148
52g/mL
34° to 40°C
Balance 2, 5-isomer
95.0
Authentic
Low Melting Crystalline Solid
96%
108.14
23, 97

Chemical Identifiers

108-50-9
100.0
C6H8N2
MFCD00006148
≥95%
95.0
96%
108.14
2, 6-Dimethylpyrazine, Balance 2, 5-isomer

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Flammable solid.
Harmful if swallowed.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning