missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylpyrazine, 96%, ACROS Organics™

Chemical Identifiers

CAS 108-50-9
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006148
Chemical Name or Material 2, 6-Dimethylpyrazine, Balance 2, 5-isomer
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC116691000 SDS Thermo Fisher Scientific
AC116691000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC116690250 SDS Thermo Fisher Scientific
116690250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108-50-9
100.0
C6H8N2
MFCD00006148
≥95%
95.0
96%
108.14
2, 6-Dimethylpyrazine, Balance 2, 5-isomer

Specifications

Low Melting Crystalline Solid
100g
34° to 40°C
23, 97
100.0
C6H8N2
MFCD00006148
Authentic
≥95%
108.14
Pale yellow
52g/mL
95.0
108-50-9
96%
Glass bottle
2, 6-Dimethylpyrazine, Balance 2, 5-isomer

Safety and Handling

GHS H Statement:
Flammable solid.
Harmful if swallowed.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning