Learn More
2,6-Dimethylpiperidine, predominantly cis, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2452418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Dimethylpiperidine | |
504-03-0 | |
0.832 | |
11°C (51°F) | |
1.44 | |
50 g | |
79827 | |
nanofin, lupetidine, naniopinum, 2,6-lupetidine, lupetidin, nanophin, nanophine, nanophyn, piperidine, 2,6-dimethyl, naniopine | |
CC1CCCC(N1)C | |
113.204 | |
113.2 |
predominantly cis | |
<-20°C | |
126°C to 129°C | |
C7H15N | |
MFCD00066304 | |
UN2733 | |
Air Sensitive | |
SDGKUVSVPIIUCF-UHFFFAOYSA-N | |
2,6-dimethylpiperidine | |
68843 | |
99% |
Chemical Identifiers
504-03-0 | |
113.204 | |
SDGKUVSVPIIUCF-UHFFFAOYSA-N | |
68843 | |
CC1CCCC(N1)C |
C7H15N | |
MFCD00066304 | |
nanofin, lupetidine, naniopinum, 2,6-lupetidine, lupetidin, nanophin, nanophine, nanophyn, piperidine, 2,6-dimethyl, naniopine | |
2,6-dimethylpiperidine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 207-981-6
RTECSNumber : OK5775000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only