Please login to your online account to display your discounted pricing

2,6-Dimethylpiperazine, 99%, predominantly cis, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC116670010

 View more versions of this product

Catalog No. AC116670010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 114.19
Quantity 1g
Molecular Formula C6H14N2
Chemical Name or Material 2, 6-Dimethylpiperazine
Assay Percent Range 99%
CAS 108-49-6

Specifications

Chemical Name or Material 2, 6-Dimethylpiperazine
Assay Percent Range 99%
Molecular Formula C6H14N2
Formula Weight 114.19