Please login to your online account to display your discounted pricing

2,6-Dimethylphenylmagnesium bromide, 1M solution in THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC436068000

 View more versions of this product

Catalog No. AC436068000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Formula Weight 209.37
Physical Form Solution
Quantity 800mL
Color Colorless to yellow-brown
Packaging AcroSeal™ Glass Bottle
Concentration 0.95 to 1.10M
Density 0.9980g/mL
Health Hazard 1 Danger
CAS Min % 19.93
CAS 21450-64-6
CAS Max % 23.08
CAS Min % 76.92
CAS Max % 80.07
CAS 109-99-9
Chemical Name or Material 2, 6-Dimethylphenylmagnesium bromide
Molecular Formula C8H9BrMg

Specifications

Chemical Name or Material 2, 6-Dimethylphenylmagnesium bromide
CAS 21450-64-6
CAS Min % 19.93
CAS Max % 23.08
CAS 109-99-9
CAS Min % 76.92
CAS Max % 80.07
Molecular Formula C8H9BrMg
Formula Weight 209.37
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.