Please login to your online account to display your discounted pricing

2,6-Dimethylphenylmagnesium bromide, 1M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dimethylphenylmagnesium bromide
CAS 21450-64-6
CAS Min % 19.93
CAS Max % 23.08
CAS 109-99-9
CAS Min % 76.92
CAS Max % 80.07
Molecular Formula C8H9BrMg
Formula Weight 209.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC436061000

 
thermo fisher scientific
AC436061000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436068000

 
thermo fisher scientific
AC436068000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 209.37
Quantity 100mL
Color Colorless to yellow-brown
Packaging AcroSeal™ Glass Bottle
Concentration 0.95 to 1.10M
Density 0.9980g/mL
CAS 21450-64-6
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face
Chemical Name or Material 2, 6-Dimethylphenylmagnesium bromide
CAS Min % 19.93
CAS 109-99-9
CAS Min % 76.92
Molecular Formula C8H9BrMg
CAS Max % 23.08
CAS Max % 80.07
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides.
Safety and Handling

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face

GHS P Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides.