Learn More
2,6-Dimethylphenylboronic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 359270010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 6-Dimethylphenylboronic acid | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD01009693 | |
2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za | |
ZXDTWWZIHJEZOG-UHFFFAOYSA-N | |
(2,6-dimethylphenyl)boronic acid | |
583322 | |
98% |
100379-00-8 | |
100.0 | |
97.5% min. (HPLC) | |
C8H11BO2 | |
1g | |
Solubility in water: .. Other solubilities: soluble in methanol | |
B(C1=C(C=CC=C1C)C)(O)O | |
149.99 | |
149.99 |
Chemical Identifiers
100379-00-8 | |
149.99 | |
ZXDTWWZIHJEZOG-UHFFFAOYSA-N | |
583322 | |
B(C1=C(C=CC=C1C)C)(O)O |
C8H11BO2 | |
MFCD01009693 | |
2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za | |
(2,6-dimethylphenyl)boronic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
RUO â Research Use Only