Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylphenyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 2,6-Dimethylphenyl isothiocyanate
CAS 19241-16-8
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD00038715
UN Number UN2922
Beilstein 637195
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1948006

  SDS
alfa aesar™
A1948006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1948014

  SDS
alfa aesar™
A1948014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1948022

  SDS
alfa aesar™
A1948022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.24
Chemical Name or Material 2,6-Dimethylphenyl isothiocyanate
Assay 98%
Molecular Formula C9H9NS
Boiling Point 247°C
MDL Number MFCD00038715
CAS 19241-16-8
UN Number UN2922
Beilstein 637195
Quantity 5g
Flash Point >110°C (230°F)
Odor Pungent
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Applications
2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.