missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylphenyl isocyanide

Manufacturer:  Alfa Aesar™ 2074803

 View more versions of this product

Catalog No. AA2074803



Specifications

Specifications

2,6-Dimethylphenyl isocyanide
73°C to 74°C
2769-71-3
MFCD00013479
2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide
CC1=C(C(=CC=C1)C)[N+]#[C-]
131.178
131.18
Unpleasant
1g
C9H9N
UN2811
DNJLFZHMJDSJFN-UHFFFAOYSA-N
2-isocyano-1,3-dimethylbenzene
76009
Crystalline

Chemical Identifiers

2769-71-3
131.178
DNJLFZHMJDSJFN-UHFFFAOYSA-N
76009
CC1=C(C(=CC=C1)C)[N+]#[C-]
C9H9N
MFCD00013479
2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide
2-isocyano-1,3-dimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents