Learn More
2,6-Dimethylphenol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC116570025
Specifications
2, 6-Dimethylphenol | |
73°C | |
Glass bottle | |
0.05% max. | |
Brown-Yellow to Colorless | |
2.5kg | |
98.5 | |
98.5% min. (GC) | |
(CH3)2C6H3OH | |
06, 485 | |
1-hydroxy-2,6-dimethylbenzene, 2,6-dimethyl phenol, 2,6-dimethyl-phenol, 2,6-dmp, 2,6-xylenol, 2-hydroxy-1,3-dimethylbenzene, 2-hydroxy-m-xylene, phenol, 2,6-dimethyl, vic-m-xylenol, xylenol 235 | |
NXXYKOUNUYWIHA-UHFFFAOYSA-N | |
2,6-dimethylphenol | |
11335 | |
Low Melting Solid |
1.1500g/mL | |
Authentic | |
1.15 | |
203.0°C | |
45.0°C to 48.0°C | |
576-26-1 | |
100.0 | |
C8H10O | |
MFCD00002240 | |
15, 10279 | |
Solubility in water: 10g/L (20°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene | |
CC1=C(C(=CC=C1)C)O | |
122.17 | |
122.17 | |
99% |
Chemical Identifiers
576-26-1 | |
122.17 | |
NXXYKOUNUYWIHA-UHFFFAOYSA-N | |
11335 | |
CC1=C(C(=CC=C1)C)O |
C8H10O | |
MFCD00002240 | |
1-hydroxy-2,6-dimethylbenzene, 2,6-dimethyl phenol, 2,6-dimethyl-phenol, 2,6-dmp, 2,6-xylenol, 2-hydroxy-1,3-dimethylbenzene, 2-hydroxy-m-xylene, phenol, 2,6-dimethyl, vic-m-xylenol, xylenol 235 | |
2,6-dimethylphenol |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-400-1
Recommended Storage : this product gradually takes up color on storage