Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylmorpholine, cis + trans, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Dimethylmorpholine
CAS 141-91-3
Name Note cis + trans
Molecular Formula C6H13NO
Formula Weight 115.18
MDL Number MFCD00005973
UN Number UN1992
Beilstein 103036
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2595414

  SDS
alfa aesar™
H2595414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2595422

  SDS
alfa aesar™
H2595422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.18
Chemical Name or Material 2,6-Dimethylmorpholine
TSCA TSCA
Name Note cis + trans
Assay 97%
Molecular Formula C6H13NO
Melting Point −85°C
Boiling Point 147°C
MDL Number MFCD00005973
CAS 141-91-3
UN Number UN1992
Beilstein 103036
Quantity 25g
Flash Point 48°C (120°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H226-H312
Causes serious eye damage.
Flammable liquid and vapour.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
RTECS Number QE1750000
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H226-H312
Causes serious eye damage.
Flammable liquid and vapour.
Harmful in contact with skin.