missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylmorpholine, 97%, mixture of isomers, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC155341000

 View more versions of this product

Catalog No. AC155341000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dimethylmorpholine
48°C
1.447
147.0°C
100g
96% min. (mixture of isomers) (GC)
2,6-dimethyl morpholine
CC1CNCC(O1)C
115.17
115.17
0.9350g/mL
Glass bottle
0.935
-85.0°C
141-91-3
C6H13NO
HNVIQLPOGUDBSU-UHFFFAOYSA-N
2,6-dimethylmorpholine
110862
97%

Chemical Identifiers

141-91-3
115.17
2,6-dimethyl morpholine
2,6-dimethylmorpholine
C6H13NO
HNVIQLPOGUDBSU-UHFFFAOYSA-N
110862
CC1CNCC(O1)C
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 205-509-3

SDS Documents