Please login to your online account to display your discounted pricing

2,6-Dimethylmorpholine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 6-Dimethylmorpholine
Assay Percent Range 97%
Molecular Formula C6H13NO
Formula Weight 115.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC155341000

  SDS
thermo fisher scientific
AC155341000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155340250

  SDS
thermo fisher scientific
AC155340250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2, 6-Dimethylmorpholine
Quantity 100g
CAS 141-91-3
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H13NO
Formula Weight 115.17