Learn More
2,6-Dimethylmorpholine, 97%, mixture of isomers, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 155340250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 6-Dimethylmorpholine | |
0.9350g/mL | |
48°C | |
Glass bottle | |
1.447 | |
0.935 | |
HNVIQLPOGUDBSU-UHFFFAOYSA-N | |
2,6-dimethylmorpholine | |
110862 | |
97% |
141-91-3 | |
147.0°C | |
96% min. (mixture of isomers) (GC) | |
C6H13NO | |
25g | |
morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod | |
CC1CNCC(O1)C | |
115.17 | |
115.17 |
Chemical Identifiers
141-91-3 | |
115.17 | |
morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod | |
2,6-dimethylmorpholine |
C6H13NO | |
HNVIQLPOGUDBSU-UHFFFAOYSA-N | |
110862 | |
CC1CNCC(O1)C |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 205-509-3
RUO â Research Use Only