Please login to your online account to display your discounted pricing

2,6-Dimethylcyclohexanone, Mixture of Isomers 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  146700250

 View more versions of this product

Catalog No. AC146700250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 126.2
Quantity 25g
Color Clear colorless to yellow
EINECS Number 220-570-6
TSCA TSCA
Density 0.9200g/mL
Beilstein 07, 23
CAS Max % 100.0
Chemical Name or Material 2, 6-Dimethylcyclohexanone
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H14O
Linear Formula (CH3)2C6H8(=O)
MDL Number MFCD00001637
CAS 2816-57-1
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)

Specifications

Chemical Name or Material 2, 6-Dimethylcyclohexanone
Name Note 99%
CAS 2816-57-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H14O
Linear Formula (CH3)2C6H8(=O)
Formula Weight 126.2
MDL Number MFCD00001637
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents