Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylcyclohexanone, cis + trans, 99%

Chemical Identifiers

Chemical Name or Material 2,6-Dimethylcyclohexanone
CAS 2816-57-1
Name Note cis + trans
Molecular Formula C8H14O
Formula Weight 126.2
MDL Number MFCD00001637
UN Number UN1224
Beilstein 1099000
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1264722

 
alfa aesar™
A1264722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1264714

 
alfa aesar™
A1264714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1264736

 
alfa aesar™
A1264736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.2
Chemical Name or Material 2,6-Dimethylcyclohexanone
Name Note cis + trans
Assay 99%
Molecular Formula C8H14O
Boiling Point 174° to 176°C
MDL Number MFCD00001637
CAS 2816-57-1
UN Number UN1224
Beilstein 1099000
Quantity 100g
Flash Point 51°C (123°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.